Strontium hydroxide octahydrate ≥99.995%

Danger

463752-5G 463752-25G 463752-100G
220069-940EA 107.54 USD
220069-940 220069-939 220069-938
Strontium hydroxide octahydrate ≥99.995%
Strontium hydroxide octahydrate
Formula: H₂O₂Sr·8H₂O
MW: 265.76 g/mol
Melting Pt: 375 °C
Density: 1.9 g/cm³ (20 °C)
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR