1-Boc-4-piperidone

Synonyms: 4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone

461350-100G 461350-25G 461350-5G
220069-854EA 554.62 USD
220069-854 220069-855 220069-856
1-Boc-4-piperidone
1-Boc-4-piperidone
Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD00151800
CAS Number: 79099-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR