p-Toluidine ≥99%

Danger

Synonyms: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine

461121-250G 461121-1KG
220069-829EA 40.92 USD
220069-829 220069-828
p-Toluidine ≥99%
p-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: 43 °C
Density: 1.046 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007906
CAS Number: 106-49-0
EINECS: 203-403-1
UN: 3451
ADR: 6.1,II
Merck Index: 12,09674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR