2,5-Dibromo-1,4-dimethoxybenzene ≥97%

Warning

Synonyms: 1,4-Dibromo-2,5-dimethoxybenzene

461105-25G 461105-5G
220069-825EA 183.91 USD
220069-825 220069-826
2,5-Dibromo-1,4-dimethoxybenzene ≥97%
2,5-Dibromo-1,4-dimethoxybenzene
Formula: C₈H₈Br₂O₂
MW: 295.96 g/mol
MDL Number: MFCD00025694
CAS Number: 2674-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR