1-Ethyl-3-methylimidazoliumhexafluorophosphate

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

46093-50G-F 46093-5G-F
220069-800EA 1228.51 USD
220069-800 220069-801
1-Ethyl-3-methylimidazoliumhexafluorophosphate
1-Ethyl-3-methylimidazoliumhexafluorophosphate
Formula: C₆H₁₁F₆N₂P
MW: 256.13 g/mol
MDL Number: MFCD00216703
CAS Number: 155371-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR