Platinium(IV) oxide

Warning

Synonyms: Platinum dioxide, Adam's type catalyst

459925-250MG 459925-1G 459925-5G
220069-632EA 60.92 USD
220069-632 220069-631 220069-633
Platinium(IV) oxide
Platinium(IV) oxide
Formula: O₂Pt
MW: 227.08 g/mol
Melting Pt: 450 °C
Density: 11.8 g/cm³ (20 °C)
MDL Number: MFCD00011184
CAS Number: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR