2,4-Dibromo-1-fluorobenzene ≥90%

Synonyms: 1,3-Dibromo-4-fluorobenzene, 1-Fluoro-2,4-dibromobenzene

459518-25ML 459518-5ML
220069-363EA 137.31 USD
220069-363 220069-364
2,4-Dibromo-1-fluorobenzene ≥90%
2,4-Dibromo-1-fluorobenzene
Formula: C₆H₃Br₂F
MW: 253.9 g/mol
MDL Number: MFCD00000283
CAS Number: 1435-53-6
EINECS: 215-860-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR