Tetramethylammonium fluoride

459135-1G 459135-5G
220069-602EA 114.15 USD
220069-602 220069-603
Tetramethylammonium fluoride
Tetramethylammonium fluoride
Formula: C₄H₁₂FN
MW: 93.14 g/mol
Melting Pt: 319 °C
Density: 1.058 g/cm³ (20 °C)
CAS Number: 373-68-2
EINECS: 206-769-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR