Zinc chloride, anhydrous

Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

456845-25G 456845-5G
220069-000EA 498.46 USD
220069-000 220069-001
Zinc chloride, anhydrous
Zinc chloride
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR