Sodium formate ≥99.998%

Synonyms: Formic acid sodium salt

456020-25G 456020-5G
220067-241EA 258.46 USD
220067-241 220069-145
Sodium formate ≥99.998%
Sodium formate
Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 257.31 °C
Density: 1.919 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 13,08694

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR