Potassium hexacyanoferrate(II) trihydrate ≥99.99%

455989-100G 455989-25G
220067-236EA 766.15 USD
220067-236 220067-237
Potassium hexacyanoferrate(II) trihydrate ≥99.99%
Potassium hexacyanoferrate(II) trihydrate
Formula: K₄Fe(CN)₆·3H₂O
Melting Pt: 70 °C
Density: 1.889 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR