Phosphotungstic acid hydrate

Danger

Synonyms: Phosphotungstic acid hydrate, Wolframophosphoric acid hydrate, Dodecawolframophosphoric acid hydrate, 12-Tungstophosphate hydrate, Tungstophosphoric Acid Hydrate

455970-10G 455970-50G
220067-234EA 266.15 USD
220067-234 220067-235
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Formula: H₃[P(W₃O₁₀)₄]·nH₂O
MW: 2898.07 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR