Ammonium hydrogen difluoride

Danger

455830-100G 455830-25G
220068-210EA 447.69 USD
220068-210 220068-211
Ammonium hydrogen difluoride
Ammonium hydrogen difluoride
Formula: NH₄HF₂
Melting Pt: 127 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR