Potassium tetrahydroborate ≥98%

Danger

Synonyms: Potassium tetrahydridoborate, Potassium borohydride

455571-100G 455571-2KG 455571-500G
220069-106EA 75.46 USD
220069-106 220069-107 220069-108
Potassium tetrahydroborate ≥98%
Potassium tetrahydroborate
Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: 500 °C
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3,I
Merck Index: 12,07777

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR