4-Ethoxyphenylboronic acid ≥97%

Synonyms: 4-Ethoxybenzeneboronic acid, (4-Ethoxyphenyl)metaboric acid, Dihydroxy(4-ethoxyphenyl)borane

455539-10G 455539-1G
220069-100EA 59.14 USD
220069-100 220069-101
4-Ethoxyphenylboronic acid ≥97%
4-Ethoxyphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD00674028
CAS Number: 22237-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR