2-Ethoxyphenylboronic acid

455520-1G 455520-10G
220069-098EA 30.46 USD
220069-098 220069-097
2-Ethoxyphenylboronic acid
2-Ethoxyphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD00674027
CAS Number: 213211-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR