α,α,α,5-Tetrafluoro-m-toluic acid ≥98%

Synonyms: 3-Fluoro-5-trifluoromethylbenzoic acid, 3-Fluoro-5-(trifluoromethyl)benzoic acid

455326-1G 455326-5G
220067-699EA 45.23 USD
220067-699 220067-700
α,α,α,5-Tetrafluoro-m-toluic acid ≥98%
α,α,α,5-Tetrafluoro-m-toluic acid
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
MDL Number: MFCD00061293
CAS Number: 161622-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR