2-(Diphenylphosphino)benzoic acid ≥97%

454885-1G 454885-5G
220068-916EA 141.92 USD
220068-916 220068-917
2-(Diphenylphosphino)benzoic acid ≥97%
2-(Diphenylphosphino)benzoic acid
Formula: C₁₉H₁₅O₂P
MW: 306.3 g/mol
MDL Number: MFCD00674024
CAS Number: 17261-28-8
EINECS: 241-293-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR