4-Nitro-2-picoline-N-oxide

Synonyms: 2-Methyl-4-nitropyridine-1-oxide, 4-Nitro-2-picoline N-oxide, 2-Methyl-4-nitropyridine-N-oxide

454850-25G 454850-5G
220068-910EA 399.54 USD
220068-910 220068-911
4-Nitro-2-picoline-N-oxide
4-Nitro-2-picoline-N-oxide
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00014625
CAS Number: 5470-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR