4-Iodo-o-toluidine ≥97%

Synonyms: 2-Amino-5-iodotoluene, 4-Iodo-2-methylaniline

454788-1G 454788-10G
220068-533EA 67.08 USD
220068-533 220068-532
4-Iodo-o-toluidine ≥97%
4-Iodo-o-toluidine
Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 91…92 °C
MDL Number: MFCD00025299
CAS Number: 13194-68-8
EINECS: 236-154-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR