Trimethyl borate ≥98%

Danger

452920-2L 452920-100ML 452920-500ML
220068-744EA 137.08 USD
220068-744 220068-743 220068-745
Trimethyl borate ≥98%
Trimethyl borate
Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.915 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3,II
Merck Index: 12,09842

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR