1-Iodo-3,5-dinitrobenzene ≥98%

452769-5G 452769-1G
220068-504EA 441.6 USD
220068-504 220068-503
1-Iodo-3,5-dinitrobenzene ≥98%
1-Iodo-3,5-dinitrobenzene
Formula: C₆H₃IN₂O₄
MW: 294.01 g/mol
MDL Number: MFCD00024330
CAS Number: 6276-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a8b0ae913903fd151f348544f806e3572bda598&vid=c5bbbe4acc4e317b7abb0b2bf5b7e073185cf3b0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR