n-Decane-[D₂₂]

Danger

452416-1G 452416-5G
220067-216EA 290.77 USD
220067-216 220067-217
n-Decane-[D₂₂]
n-Decane-[D₂₂]
Formula: D₃C(CD₂)₈CD₃
CAS Number: 16416-29-8
UN: 2247
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR