Tellurium ≥99.999%

Danger

452378-25G 452378-5G
220067-210EA 664.62 USD
220067-210 220067-211
Tellurium ≥99.999%
Tellurium
Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR