Antimony ≥99.999%

Warning

452343-50G 452343-10G
220067-207EA 875.38 USD
220067-207 220067-206
Antimony ≥99.999%
Antimony
Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR