Iron(III) chloride, anhydrous

Danger

451649-1G 451649-5G
220068-617EA 207.69 USD
220068-617 220068-676
Iron(III) chloride, anhydrous
Iron(III) chloride
Formula: FeCl₃
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 260 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8,III
Merck Index: 13,04048

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR