Lithium tetrafluoroborate, anhydrous ≥99.998%

451622-25G 451622-5G
220068-614EA 906.15 USD
220068-614 220068-615
Lithium tetrafluoroborate, anhydrous ≥99.998%
Lithium tetrafluoroborate
Formula: BF₄Li
MW: 93.74 g/mol
Density: 0.852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR