Nickel(II) chloride, anhydrous

Danger

Synonyms: Nickel dichloride

451193-25G 451193-5G
220067-677EA 1158.46 USD
220067-677 220067-678
Nickel(II) chloride, anhydrous
Nickel(II) chloride
Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1009 °C
Density: 3.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 12,06588

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR