Lutetium trichloride, anhydrous ≥99.99%

450960-1G 450960-5G
220068-597EA 249.23 USD
220068-597 220068-598
Lutetium trichloride, anhydrous ≥99.99%
Lutetium trichloride
Formula: Cl₃Lu
MW: 281.33 g/mol
Melting Pt: 892 °C
Density: 3.98 g/cm³ (25 °C)
MDL Number: MFCD00011099
CAS Number: 10099-66-8
EINECS: 233-240-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR