Gallium(III) chloride, anhydrous ≥99.99%

Synonyms: Gallium trichloride

450898-50G 450898-5G 450898-25G
220068-587EA 441.54 USD
220068-587 220068-588 220068-586
Gallium(III) chloride, anhydrous ≥99.99%
Gallium(III) chloride
Formula: Cl₃Ga
MW: 176.08 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 77.9 °C
Density: 2.29 g/cm³ (80 °C)
MDL Number: MFCD00011018
CAS Number: 13450-90-3
EINECS: 236-610-0
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR