Rhodium(III) chloride hydrate

Synonyms: Rhodium trichloride hydrate

450286-250MG 450286-1G
220068-299EA 404.62 USD
220068-299 220068-298
Rhodium(III) chloride hydrate
Rhodium (III) chloride hydrate
Formula: Cl₃Rh·H₂O
MW: 227.28 g/mol
MDL Number: MFCD00149839
CAS Number: 20765-98-4
EINECS: 233-165-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR