Silver(I) hexafluorophosphate

Synonyms: Silver hexafluorophosphate

450219-5G 450219-1G
220068-287EA 427.69 USD
220068-287 220068-286
Silver(I) hexafluorophosphate
Silver(I) hexafluorophosphate
Formula: AgF₆P
MW: 252.83 g/mol
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR