Lithium acetate dihydrate ≥99.999%

450189-5G 450189-25G
220068-282EA 441.54 USD
220068-282 220068-281
Lithium acetate dihydrate ≥99.999%
Lithium acetate dihydrate
Formula: H₃CCOOLi·2H₂O
Melting Pt: 291 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR