Barium hydroxide hydrate ≥99.995%

Danger

450170-25G 450170-5G
220068-279EA 267.69 USD
220068-279 220068-280
Barium hydroxide hydrate ≥99.995%
Barium hydroxide hydrate
Formula: H₂BaO₂·H₂O
MW: 189.36 g/mol
Storage Temperature: Ambient
CAS Number: 40226-30-0
EINECS: 241-234-5
UN: 3262
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR