Calcium hydroxide ≥99.995%

Danger

450146-25G 450146-5G
220068-274EA 463.08 USD
220068-274 220068-275
Calcium hydroxide ≥99.995%
Calcium hydroxide
Formula: Ca(OH)₂
Melting Pt: 550 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III
Merck Index: 13,01675

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR