Yttrium(III) chloride, anhydrous

Warning

450103-5G 450103-25G
220068-537EA 184.62 USD
220068-537 220068-492
Yttrium(III) chloride, anhydrous
Yttrium(III) chloride
Formula: Cl₃Y
MW: 195.26 g/mol
Boiling Pt: 1488 °C (1013 hPa)
Melting Pt: 703 °C
Density: 2.52 g/cm³ (700 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011470
CAS Number: 10361-92-9
EINECS: 233-801-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR