Ytterbium(III) chloride, anhydrous ≥99.99%

Warning

450073-5G 450073-25G
220068-491EA 289.23 USD
220068-491 220068-490
Ytterbium(III) chloride, anhydrous ≥99.99%
Ytterbium(III) chloride
Formula: Cl₃Yb
MW: 279.4 g/mol
Melting Pt: 860 °C
MDL Number: MFCD00011288
CAS Number: 10361-91-8
EINECS: 233-800-5
Merck Index: 13,10014

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR