Strontium fluoride, anhydrous ≥99.99%

Warning

450030-10G 450030-50G
220068-488EA 193.85 USD
220068-488 220068-489
Strontium fluoride, anhydrous ≥99.99%
Strontium fluoride
Formula: F₂Sr
MW: 125.62 g/mol
Boiling Pt: 2460 °C (1013 hPa)
Melting Pt: 1477 °C
Density: 4.24 g/cm³ (20 °C)
MDL Number: MFCD00011251
CAS Number: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III
Merck Index: 12,09002

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR