Neodymium trichloride, anhydrous ≥99.99%

Warning

Synonyms: Neodymium (III) chloride

449946-25G 449946-5G
220068-474EA 1692.31 USD
220068-474 220068-475
Neodymium trichloride, anhydrous ≥99.99%
Neodymium trichloride
Formula: Cl₃Nd
MW: 250.6 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 758 °C
Density: 4.13 g/cm³ (20 °C)
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR