Lithium fluoride, anhydrous ≥99.99%

Warning

449903-2G 449903-10G
220068-469EA 141.08 USD
220068-469 220068-468
Lithium fluoride, anhydrous ≥99.99%
Lithium fluoride
Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676 °C (1013 hPa)
Melting Pt: 870 °C
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,05553

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR