Lithium chloride, anhydrous ≥99.9%

Warning

449881-50G 449881-10G
220068-467EA 410.77 USD
220068-467 220068-466
Lithium chloride, anhydrous ≥99.9%
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR