Lanthanum(III) fluoride, anhydrous ≥99.99%

Synonyms: Lanthanum trifluoride

449857-25G 449857-5G
220068-182EA 787.69 USD
220068-182 220068-183
Lanthanum(III) fluoride, anhydrous ≥99.99%
Lanthanum(III) fluoride
Formula: F₃La
MW: 195.9 g/mol
Boiling Pt: ∼2327 °C (1013 hPa)
Melting Pt: 1491.85…1507.85 °C
Density: 5.936 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS Number: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR