Lanthanum(III) chloride, anhydrous ≥99.99%

Warning

Synonyms: Lanthanum chloride, Lanthanum trichloride

449830-25G 449830-5G
220068-180EA 500 USD
220068-180 220068-181
Lanthanum(III) chloride, anhydrous ≥99.99%
Lanthanum(III) chloride
Formula: Cl₃La
MW: 245.26 g/mol
Melting Pt: 857 °C
Density: 3.8773 g/cm³ (-0.15 °C)
MDL Number: MFCD00011068
CAS Number: 10099-58-8
EINECS: 233-237-5
Merck Index: 13,05379

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR