Erbium trichloride, anhydrous ≥99.9%

Warning

449792-5G 449792-25G
220068-171EA 296.92 USD
220068-171 220068-170
Erbium trichloride, anhydrous ≥99.9%
Erbium trichloride
Formula: Cl₃Er
MW: 273.62 g/mol
Boiling Pt: 1500 °C (1013 hPa)
Melting Pt: 751 °C
Density: 4.1 g/cm³ (20 °C)
MDL Number: MFCD00010989
CAS Number: 10138-41-7
EINECS: 233-385-0

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR