Trioctylsilane

Warning

448923-25ML 448923-5ML
220068-396EA 279.23 USD
220068-396 220068-397
Trioctylsilane
Trioctylsilane
Formula: C₂₄H₅₂Si
MW: 368.76 g/mol
MDL Number: MFCD00053812
CAS Number: 18765-09-8
EINECS: 242-559-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR