2-Bromo-3-methylpyridine

Warning

448109-25ML 448109-5ML
220068-267EA 205.69 USD
220068-267 220068-268
2-Bromo-3-methylpyridine
2-Bromo-3-methylpyridine
Formula: C₆H₆BrN
MDL Number: MFCD00239380
CAS Number: 3430-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR