(±)-3-Butyne-2-ol ≥97%

Synonyms: (±)-But-3-yn-2-ol, (±)-3-Hydroxy-1-butyne

447986-25ML 447986-100ML
220068-263EA 219.69 USD
220068-263 220068-262
(±)-3-Butyne-2-ol ≥97%
(±)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 104 - 108 °C (1013 hPa)
Melting Pt: -12 °C
Density: 0.895 g/cm³ (20 °C)
MDL Number: MFCD00004541
CAS Number: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR