5-Chlorosalicylaldehyde ≥98%

Warning

Synonyms: 5-Chloro-2-hydroxybenzaldehyde

447706-25G 447706-5G
220066-532EA 76.89 USD
220066-532 220066-533
5-Chlorosalicylaldehyde ≥98%
5-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 105 °C (12 -rr)
Melting Pt: 100.3 °C
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR