3,5-Di-tert-butyl-4-hydroxybenzyl alcohol ≥97%

446424-5G 446424-25G
220068-136EA 55.89 USD
220068-136 220068-135
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol ≥97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
MDL Number: MFCD00017254
CAS Number: 88-26-6
EINECS: 201-815-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR