2-Methoxyphenylboronic acid ≥95%

Synonyms: 2-Methoxybenzeneboronic acid, (2-Methoxyphenyl)metaboric acid, Dihydroxy(2-methoxyphenyl)borane

445231-1G 445231-5G
220067-988EA 30.18 USD
220067-988 220067-989
2-Methoxyphenylboronic acid ≥95%
2-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00236047
CAS Number: 5720-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR